Sulfathiazole

Supplier: LGC Standards PROMOCHEM

Synonyms: 4-Amino-N-(thiazol-2-yl)benzenesulphonamideN1-2-Thiazolylsulphanilamid

PROOMM0578.00EA 290 EUR
PROOMM0578.00
Sulfathiazole
Sulfathiazole
Formula: C₉H₉N₃O₂S₂
MW: 255,32 g/mol
MDL Number: MFCD00005319
CAS Number: 72-14-0
EINECS: 200-771-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR