Fixed stainless steel probe (25,4 mm)
- Fast, one second response for measuring ambient air temperature
- Designed for penetration of semisolids and liquids
- Accurate readings when immersed only 8,5 mm
- Provides readings in °F and °C
This lightweight, pocket-sized thermometer can be easily carried. Thanks to its attached key chain, users can clip it to a lab coat buttonhole, jacket zipper, or their keys. It can also be hung in storerooms or fume hoods for easy monitoring.
Certifications: A serial numbered certificate is provided from an ISO/IEC 17025:2005 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Delivery information: Supplied with a key chain, a probe, a battery powers unit for one year of continuous operation, and a Traceable® calibration certificate.