Micro magnet for glass pumps, BOLA
Supplier: Bohlender
BOLA
BOHLG876-14EA 148 EUR
BOHLG876-14
Micro magnet for glass pumps, BOLA
Magnets
Made of PTFE, the magnet can be adjusted in height by means of the integrated screw. The strong magnet provides good power transmission. Suitable for all common laboratory glass pumps, e.g. from company NORMAG®.
  • Chemical resistance
  • Temperature resistance from +0 to +150 °C