Bis-acrylamide powder, Bis-2™, Ultra Pure Grade
0832-500MLEA 29 EUR
VWRC0832-500ML
Bis-acrylamide powder, Bis-2™, Ultra Pure Grade
Acrylamide and bis-acrylamide pre-weighed powder blends, pre-mixed stock solutions, and ready to use solutions for customised polyacrylamide gel electrophoresis of proteins and PAGE of nucleic acids.
  • Ultra-pure powder with acrylamide purity >99,9%
  • Convenient, ready to use
  • Eliminates hazardous powdered acrylamide handling