Sodium carbonate, Reagent Grade

Supplier: VWR Chemicals
Warning

0246-1KGEA 130 EUR
0246-1KG
Sodium carbonate, Reagent Grade
Sodium carbonate
Formula: Na₂CO₃
MW: 105,99 g/mol
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2,53 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,08596
https://ie.vwr.com/stibo/web/std.lang.all/05/89/16640589.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR