Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (5)
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

You Searched for: "IN/Share"

https://ie.vwr.com/stibo/smallweb/std.lang.all/48/84/18944884.jpg
SearchPresentationType-VERTICAL
 
 
SearchResultCount:"33"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Merck
Description:   Ready-to-use solution for the preparation of polyacrylamide gels.
UOM:  1 * 1 l
Supplier:  VWR Chemicals
Description:   Premixed, ready to pour acrylamide solution and 20X running buffer for the SDS PAGE analysis of proteins.
Supplier:  VWR Chemicals
Description:   High quality standard agarose for routine analysis with broad separation range (0,05 kbp to 50 kbp). Ideal for blotting applications due to high gel s...
Supplier:  VWR Chemicals
Description:   Acrylamide-​Bis solution (37.5: 1) 30% aqueous solution
UOM:  1 * 500 ml
Supplier:  VWR Chemicals
Description:   Agarose 3:1 HRB™ (High Resolution Blend) is a mixture of agarose formulated to provide high resolution of small nucleic acids and PCR products. This p...
Supplier:  VWR Chemicals
Description:   Acryl/​Bis solution (30%) 29:1
UOM:  1 * 500 ml
Catalog Number: (K677-100G)
Supplier:  VWR Chemicals
Description:   Agarose I™/TBE and Agarose I™/TAE powder blends have TBE or TAE buffer already mixed in with agarose at a 1X concentration. This product is not only e...
UOM:  1 * 100 g
Catalog Number: (17-1310-01)
Supplier:  GE Healthcare - Life Sciences
Description:   PlusOne™ Acrylamide IEF is outstanding for use in isoelectric focusing and DNA sequencing. In addition to the absence of impurities that interfere wit...
UOM:  1 * 1 l
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 8  of 33
  1  2  3  4  5  Next