Sample bottles, wide neck, with cap, WHEATON®
Supplier: DWK Life Sciences
WHEA990476EA 308 EUR
WHEA990476
Sample bottles, wide neck, with cap, WHEATON®
Bottles Glass Bottles
Borosilicate glass, clear.
  • For use with Sub Surface Grab Sampler III
  • 70-400 screw cap with PTFE liner
  • Autoclavable
These wide mouth sample bottles can be repeatedly autoclaved and come with PTFE-lined caps.