Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
  • Supplier
  • Refine by Suppliers
    Sort by:

  • Search Within Results

prepared-media-in-bags-powder

SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
 
 
SearchResultCount:"17"
  List View Grid View Easy View (new)
Sort by:
 
 
 
 

Supplier:  Merck
Description:   Readycult® Coliform and Readycult® Enterococci make it easy to detect coliforms, E. coli or faecal streptococci in water. You only need a steri...
Supplier:  Merck
Description:   Readybag® gamma-irradiated and pre-weighed granulated media pouches for fast, flexible and easy pathogen testing. Readybag® media pouches speed up and...
Supplier:  VWR Chemicals
Description:   MediaBag, BPW, 1530 g, Media-Bag™, Application: Enrichment, non selective, For: 60000 ml
Supplier:  VWR Chemicals
Description:   For the preliminary, non selective enrichment of bacteria, particularly pathogenic enterobacteriaceae, from foodstuffs.
Supplier:  VWR Chemicals
Description:   Each bag contains a pre-weighed amount of granulated media in a double bagged bag and is irradiated. Just in time enrichment - add sterile water and s...
Catalog Number: (7101.00450)
Supplier:  VWR Chemicals
Description:   MediaBags are filled with pre-weighed granulated media in a double bagged bag and irradiated. Add sterile water under aseptic conditions.
UOM:  1 * 100 items
Supplier:  VWR Chemicals
Description:   For the preliminary, non selective enrichment of bacteria.
Supplier:  VWR Chemicals
Description:   MediaBags are filled with pre-weighed granulated media in a double bagged bag and irradiated. Add sterile water under aseptic conditions.
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is dicontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 8  of 17
  1  2  3  Next