2-Amino-2-cyanoacetamide for synthesis, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

8.14902.0005EA 0 EUR
8.14902.0005 8.14902.0050
2-Amino-2-cyanoacetamide for synthesis, Sigma-Aldrich®
2-Amino-2-cyanoacetamide
Formula: C₃H₅N₃O
MW: 99,0913 g/mol
MDL Number: MFCD00040532
CAS Number: 6719-21-7
EINECS: 229-771-3

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point 119 - 122 °C
Molar Mass 99.09 g/mol

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR