VWR®, Chicken lysozyme (Muramidase) (from egg white)
Supplier: VWR
0663-5GEA 186 EUR
VWRC0663-5G
VWR®, Chicken lysozyme (Muramidase) (from egg white)
A hydrolytic enzyme specific for proteins found in the lipid bilayer of bacteria. It is useful for lysing gram positive and gram negative bacteria for subsequent nucleic acid extraction.
  • White crystallised powder
  • Store it frozen