Would you like to log in again?
Rescue hoist, Miller® Manhandler MN20
Supplier: HONEYWELL SAFETY PRODUCTS
Miller® Manhandler
DALL1005042EA 1220 EUR InStock
DALL1005042
Rescue hoist, Miller® Manhandler MN20
Fall Protection Fall Arresters
This compact and robust Manhandler with zinc-plated steel rope is both a rescue hoist and a load hoist.
  • Integrated attachments for MN10 tripod
  • Complete with a pulley, overspeed brake and fall indicator
  • Easy to handle
  • Transmission ratio 5:1
  • Maximum workload: 159 kg
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty