Earmuffs, Leightning® L0F
Supplier: Howard Leight by Honeywell
DALL1018010EA 27.2 EUR
DALL1018010
Earmuffs, Leightning® L0F
Ultra slim earmuffs with patented Air Flow Control™ technology delivers optimal attenuation across all frequencies, without increasing earcup size or weight. A patented baseplate chamber and high-tech non-woven layer manage the flow of air inside the earmuff to control how sound reaches the ear.
  • Robust steel headband withstands demanding use, especially in tough environments
  • Padded foam headband delivers long-wearing comfort with minimal pressure on the head
  • Slim cups and foldable headband with 'camouflage' pattern
  • Snap-in ear cushions make replacement quick and easy
For all industrial noise environments.
SNR (dB): 25