Polysafe cover spectacles
Supplier: Honeywell Safety
Polysafe
DALL1002549EA 60.3 EUR
DALL1002549
Polysafe cover spectacles
Spectacles
These cover glasses (OTG) provide a wide field of vision.
  • Highest visual quality enables clear, undistorted vision
  • Perforated side frame for fastening a safety strap
  • Single-piece polycarbonate lens with integrated side parts
  • PC lens provides 99.9% UV protection
Can be worn over prescriptive spectacles.
Weight: 40 g
Certifications: EN 166