Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safety cabinets for active storage of drums, type 90, SUPPLY line

Supplier: Düperthal
The safety storage cabinets of the SUPPLY line are ideal for efficiently and safely withdrawing flammable liquids. They have a variety of additional features which help increase safety and work comfort.

  • Ergonomic lock positioning at handle level and integrated locking cylinders in the door improve user comfort
  • One-hand operation of the easy-moving doors
  • Extraction of hazardous vapours from every cabinet level, easy visual inspection of the venting cut-off flaps
  • An exhaust air monitoring unit is integrated, as a standard, optionally with or without ventilator
  • Heavy load drawer with safety locking and maximum load bearing capacity of 250 kg facilitates loading and unloading a 200 litre drum into or from the safety storage cabinet.

The wing doors and outer carcass are made of conductive powder-coated sheet steel. Inner carcass made of high-quality decor panels. Bottom tray made of conductive, powder-coated sheet steel and galvanised grating.

Colour’s for wing doors are available optionally in gentian blue or light grey. Complete carcass in light grey.

The base design makes the cabinet easily accessible for transport from all sides. The adjustable bases have an extra- large bearing surface for easy alignment and level adjustment from inside and outside of the cabinet.

Equipotential bonding saddles at the rear of the cabinet are installed as a standard for the connection to the earthing in accordance with TRBS 2153 (prevention of ignition hazards). The interior fittings are conductively connected with the equipotential bonding saddles. Including earthing cables with crocodile clips in the cabinet.

Certifications: Conforms with DIN EN 14470-1, EK5/AK4 09-10, DIN EN 14727, DIN EN 60079-10, TRGS 510 Annex 3, TRGS 509 and GS certified.




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Description W×D×H W×D×H int. Doors Weight Pk VWR Catalog Number Quantity
Safety cabinet for active storage of drums, 1 bottom tray for 60-l drums left, 1 bottom tray for one 200-l drum right, each with grating and base, SUPPLY XXL 2 1655×745×2080 744×595×1515 2 wing doors 873 1 DUEH49-201773-047
Safety cabinet for active storage of drums, 1 bottom tray for 60-l drums left, 1 bottom tray for one 200-l drum right, each with grating and base, SUPPLY XXL 2 1655×745×2080 744×595×1515 2 wing doors 873 1 DUEH49-201773-048
Safety cabinet for active storage of drums, with 1 bottom trays for 200-l drums, 2 earthing cables and base, SUPPLY LL 2 895×745×2080 799×595×1200 2 wing doors 444 1 DUEH49-200970-038
Safety cabinet for active storage of drums, with 1 bottom trays for 200-l drums, 2 earthing cables and base, SUPPLY LL 2 895×745×2080 799×595×1200 2 wing doors 444 1 DUEH49-200970-037
Safety cabinet for active storage of drums, with 1 bottom trays for 200-l drums, 4 earthing cables and base, SUPPLY XXL 3 1655×745×2080 744×595×1515 2 wing doors 885 1 DUEH49-201773-038
Safety cabinet for active storage of drums, with 1 bottom trays for 200-l drums, 4 earthing cables and base, SUPPLY XXL 3 1655×745×2080 744×595×1515 2 wing doors 885 1 DUEH49-201773-037
Safety cabinet for active storage of drums, with 1 bottom trays for 60-l drums, 2 earthing cables and base, SUPPLY LL 1 895×745×2080 799×595×1535 2 wing doors 432 1 DUEH49-200970-018
Safety cabinet for active storage of drums, with 1 bottom trays for 60-l drums, 2 earthing cables and base, SUPPLY LL 1 895×745×2080 799×595×1535 2 wing doors 432 1 DUEH49-200970-017
Safety cabinet for active storage of drums, with 1 bottom trays for 60-l drums, 4 earthing cables and base, SUPPLY XXL 1 1655×745×2080 744×595×1515 2 wing doors 861 1 DUEH49-201773-017
Safety cabinet for active storage of drums, with 1 bottom trays for 60-l drums, 4 earthing cables and base, SUPPLY XXL 1 1655×745×2080 744×595×1515 2 wing doors 861 1 DUEH49-201773-018
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description For Pk VWR Catalog Number Quantity
Compressed air media pump set, explosion protected Safety cabinets for drums 1 DUEH50-70000-052
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty