Antipain dihydrochloride
Supplier: G-Biosciences
786-045EA 248 EUR
786-045
Antipain dihydrochloride
Proteins and Peptides
Antipain hydrochloride inhibits Ca2+-dependent endopeptidases, including papain, trypsin-like serine proteases, some cysteine proteases and to a lesser extent plasmin. It has a higher specificity for trypsin and papain compared to leupeptin.