3,3',5,5'-Tetramethylbenzidine dihydrochloride monohydrate

Supplier: Molekula

Synonyms: 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine dihydrochloride xhydrate,TMB-HCl hydrate,Dihydrochloride TMB hydrate

M27169695
MOLEM27169695EA 0 EUR
MOLEM27169695
3,3',5,5'-Tetramethylbenzidine dihydrochloride monohydrate
3,3',5,5'-Tetramethylbenzidine dihydrochloride hydrate
Formula: C₁₆H₂₀N₂·2HCl·xH₂O
MW: 313,27 g/mol (anhy)
MDL Number: MFCD00150104
CAS Number: 207738-08-7
EINECS: 264-769-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR