(Chloromethyl)triphenylphosphonium chloride 95% tech.

Warning

Synonyms: (chloromethyl)triphenylphosphoniumchloride

ACRO109530050EA 55.5 EUR
ACRO109530050 ACRO109530250
(Chloromethyl)triphenylphosphonium chloride 95% tech.
(Chloromethyl)triphenylphosphonium chloride
Formula: C₁₉H₁₇Cl₂P
MW: 347,2180 g/mol
Melting Pt: 258…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00011867
CAS Number: 5293-84-5
EINECS: 226-139-9

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 258 to 264 °C (Dec)
Titration Argentometric ≥94 %
Barium (Ba) ≤30 ppm
Calcium (Ca) ≤100 ppm
Iron (Fe) ≤10 ppm
Potassium (K) ≤100 ppm
Sodium (Na) ≤2000 ppm
Heavy metals (ICP-OES) ≤20 ppm
Titrable base ≤0.008 meq/g
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR