(2-Methylbenzyl)triphenylphosphonium chloride ≥98%
Supplier: Thermo Scientific
|
Warning
|
A10719.30EA
0
EUR
A10719.30
(2-Methylbenzyl)triphenylphosphonium chloride ≥98%
(2-Methylbenzyl)triphenylphosphonium chloride
Formula:
C₂₆H₂₄ClP MW: 402,91 g/mol Melting Pt: 270…275 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031577 CAS Number: 63368-36-5 EINECS: 264-112-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...