Potassium hydrogenarsenate dihydrate

Supplier: Molekula
Danger

MOLE54082809-5GEA 0 EUR
MOLE54082809-5G
Potassium hydrogenarsenate dihydrate
Potassium hydrogenarsenate dihydrate
Formula: HAsK₂O₄·2H₂O
CAS Number: 308103-48-2
UN: 1677
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR