Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trimethylenebis(triphenylphosphonium bromide) ≥98%

Supplier: Alfa Aesar

Purity: 98+%
CAS No.: 7333-67-7
Molecular Formula: C39H36Br2P2
Formula Weight: 726.48
Melting Point: 333-335°
Beilstein Reference: 4281936


Formula: C₃₉H₃₆Br₂P₂
CAS Number: 7333-67-7
EINECS: 230-835-8

Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 25 g B25165.14
Glass bottle for solids 5 g B25165.06
Glass bottle for solids 100 g B25165.22
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Melting point 333-335°
Formula weight 726.48
CAS No. 7333-67-7
Purity 98+%
Molecular formula C39H36Br2P2
MDL No. MFCD00051885
Beilstein reference 4281936
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Select Product:

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty