Thermal cyclers, Techne® ³Prime

Supplier: Cole Parmer

732-1608EA 4460 EUR
TECN3PRIMEG/02 TECN3PRIMEG/05 TECN3PRIMEBASE/05 TECN3PRIMEX/USB TECN3PRIMEX/02 TECN3PRIMEX/05
Thermal cyclers, Techne® ³Prime
The ³Prime personal cycler offers easy programming and instrument flexibililty, whilst maintaining the rapid ramp rates and small footprint that make it ideal for research and teaching laboratories. Using a colour touch screen and ready to go templates the user is intuitively guided to create even complicated programs, with a methodology that is standardised across the whole Techne® thermal cycler range.

  • Colour touch screen for fast program set-up
  • Fast ramp rate of up to 3,0 °C/s
  • Temperature range +4 to 100 °C
  • Gradient cycling is available directly using the ³PrimeG instrument or by upgrading the ³PrimeX unit
  • Protocols and temperature logs transferred between instruments and PC via USB

The ³PrimeX mid-sized cyclers deliver all the features of the ³Prime instrument but with an expanded sample capacity, accommodating 48×0,2 ml microtubes or 30×0,5 ml tube. Gradient cycling can be achieved directly using the ³PrimeG instrument or by upgrading the ³PrimeX unit.

The ³PrimeG is a small gradient thermal cycler that builds on all the features of the ³PrimeX instrument. The 48-well block format offers eight columns for annealing temperature optimisation and six rows for optimising reagents such as MgCl₂ and primer concentrations. Annealing temperatures can be optimised over 14 °C between temperatures 30 to 80 °C. The gradient calculator function displays the temperature for each of the eight columns, ensuring easy replication of thermal conditions.

Note: Exchange of thermal cycler blocks must be carried out by a trained service engineer. Please contact your local Avantor office for details about replacement blocks available.

* Recorded at 55 °C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR