You Searched For: Micro Valves
Micro valves are available as both screw top and crimp top vials. When used with a glass vial, only PTFE and glass are in contact with the contents. Operation is extremely simple, using the green and red buttons in tandem with a needle inserted through the septum to retrieve the sample. Micro push-bottom valves are highly dependable, leak-tight closures for vials and other laboratory containers. They provide an inert, high-pressure seal, and can be used repeatedly for unlimited syringe-needle access to sample.
Micro valves are available as both screw top and crimp top vials. When used with a glass vial, only PTFE and glass are in contact with the contents. Operation is extremely simple, using the green and red buttons in tandem with a needle inserted through the septum to retrieve the sample. Micro push-bottom valves are highly dependable, leak-tight closures for vials and other laboratory containers. They provide an inert, high-pressure seal, and can be used repeatedly for unlimited syringe-needle access to sample.
3  results were found
SearchResultCount:"3"
Sort Results
List View Easy View
Rate These Search Results
Supplier: JOUR RESEARCH (VICI)
Description: These micro valves for LC and GC has a unique design of the fitting detail which allows a leak-free seal with no potential for rotor damage from overt...
Supplier: UPCHURCH SCIENTIFIC
Description: For fine control of fluid flow rates, these valves can reduce outgoing flow. These needle valves are perfect for use with peristaltic pump fluid trans...
Catalog Number: (UPCHP-450)
Supplier: UPCHURCH SCIENTIFIC
Description: Designed to accurately split and control the low flow stream of a single incoming supply.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us on +353 1 88 22222.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at eurega_services@eu.vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organisation. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
Product(s) marked with this symbol are discontinued - sold till end of stock. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service on +353 1 8822222.
1 - 3 of 3
no targeter for Bottom