Speciality harnesses, Kevlar® 650K
Supplier: Honeywell Safety
111-1408EA 541 EUR
DALL1012338 DALL1012337 DALL1012336
Speciality harnesses, Kevlar® 650K
Heat and fire resistant Kevlar® harness, ideal for use offshore, in the Army, or by fire brigades.
  • Adjustable shoulder and leg straps
  • Clips on shoulder straps for lanyard when not in use
  • Special flame-retardant Nomex® webbing protects against weld splatter and sparks
  • Kevlar® inner core adds strength and high temperature resistance
  • Increased safety thanks to the bright contrasting colours, which facilitate inspection
EN 361, EN 358
Certifications: EN 361, EN 358