Diphenyleneiodonium chloride, Millipore®
Supplier: Merck Millipore Calbiochem
Synonyms:
Diphenyleneiodonium Chloride
300260-10EA
286
EUR
300260-10
Diphenyleneiodonium chloride, Millipore®
Diphenyleneiodonium chloride
Formula:
C₁₂H₈ClI MW: 314,55 g/mol |
MDL Number:
MFCD00214165 CAS Number: 4673-26-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...