Tri-tert-butylphosphonium tetrafluoroborate 97+%
|
Warning
|
Synonyms:
Tri-t-butylphosphonium tetrafluoroborate
ACRO366940010EA
122
EUR
ACRO366940010
ACRO366940050
Tri-tert-butylphosphonium tetrafluoroborate 97+%
Tri-tert-butylphosphonium tetrafluoroborate
Formula:
C₁₂H₂₈BF₄P MW: 290,13 g/mol Melting Pt: 261 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD04039975 CAS Number: 131274-22-1 |
Specification Test Results
Appearance | White Crystals and/or chunks |
Infrared spectrum | Conforms |
NMR (31P-NMR) | ≥97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...