Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chlorotitanium triisopropoxide 1 mol/l in hexane for synthesis

Supplier: Merck Millipore



Formula: C₉H₂₁ClO₃Ti
MW: 260,58 g/mol
MDL Number: MFCD00009861
CAS Number: 20717-86-6
UN: 1208
ADR: 3,II
Specification Test Results
Solubility (20 °C) (decomposition)
Boiling Point 68 - 70 °C (1013 hPa)
Vapor Pressure 160 hPa (20 °C)
Flash Point -22 °C
Explosion Limit 1.1 - 7.4 %(V)
Density 0.77 g/cm3 (20 °C)
Ignition Point 240 °C

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle 100 ml 8.18268.0100
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty