Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine ≥98%

Supplier: Alfa Aesar

Purity: 98%
CAS No.: 31886-57-4
Molecular Formula: C14H19FeN
Formula Weight: 257.16
Flash Point: 95°(203°F)
Refractive Index: 1.5890
Boiling Point: 120-121°/0.7mm
Optical Rotation: -13° (c=1 in ethanol, 5H)
MDL No.: MFCD00066272


Formula: C₁₄H₁₉FeN
CAS Number: 31886-57-4


 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 250 mg L19680.MD
Glass bottle for solids 1 g L19680.03
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Formula weight 257.16
CAS No. 31886-57-4
Purity 98%
Refractive index 1.5890
Flash point 95°(203°F)
Boiling point 120-121°/0.7mm
Molecular formula C14H19FeN
MDL No. MFCD00066272
Optical rotation -13° (c=1 in ethanol, 5H)
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty