Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Bromo-5-formylthiazole ≥95%

Supplier: Alfa Aesar
Warning



Formula: C₄H₂BrNOS
MW: 192,03
Melting Pt: 92…96 °C
Storage Temperature: Ambient
MDL Number: MFCD03788567
CAS Number: 464192-28-7

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 250 mg H31096.MD
Glass bottle for solids 1 g H31096.03
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Melting point 92-96°
Formula weight 192.03
CAS No. 464192-28-7
Purity 95%
Molecular formula C4H2BrNOS
MDL No. MFCD03788567
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty