Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-4-Methyl-β-nitrostyrene ≥98%

Supplier: Alfa Aesar
Warning



Formula: C₉H₉NO₂
MW: 163,18
Melting Pt: 100…104 °C
MDL Number: MFCD00024828
CAS Number: 7559-36-6

 

ORDER

Expand All / Collapse All
Pack type Pk VWR Catalog Number Quantity
Glass bottle for solids 25 g A14721.14
Glass bottle for solids 100 g A14721.22
Glass bottle for solids 5 g A14721.06
Expand All / Collapse All
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Melting point 100-104°
Formula weight 163.18
CAS No. 5153-68-4
Purity 98%
Storage temperature 2°C to 8°C
Molecular formula C9H9NO2
Beilstein reference 1864209
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty