Eriochrome cyanine R

Supplier: Molekula

Synonyms: Chromoxane Cyanine R

MOLE53361433EA 0 EUR
MOLE53361433
Eriochrome cyanine R
Eriochrome cyanine R
Formula: C₂₃H₁₅Na₃O₉S
MDL Number: MFCD00012411
CAS Number: 3564-18-9
EINECS: 222-641-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR