Would you like to log in again?
Stepsaver™ 50 v39 8 x 39 ml, titanium, 50,000 rpm/241,800 x g (with complete accessories as listed in rotor catalogue) 1 * 1 items
Supplier: SORVALL
ORDER
Expand All / Collapse All
Description VWR Catalog Number Unit Quantity
Stepsaver™ 50 v39 8 x 39 ml, titanium, 50,000 rpm/241,800 x g (with complete accessories as listed in rotor catalogue) 1 * 1 items SORV79245 1 * 1 items
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty