Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Rotors for Sorvall™ WX Ultra series

Supplier: Thermo Scientific
Sorvall®
Titanium or aluminium made rotors for WX Ultra Series centrifuges.



 

ORDER

Expand All / Collapse All
Description Pk VWR Catalog Number Quantity
Fixed angle rotor, T-1270, titanium, 12×12,5 ml tubes (16×80 mm), 448811 ×g, 70000 min-1 1 521-1963
Fixed angle rotor, T-647.5, titanium, 6×100 ml, 47500 min-1 ,250260 ×g 1 SORV08322
Swinging bucket rotor, TH-660, titanium, 6×4,4 ml, 60000 min-1, 488576 ×g 1 SORV10502
Fixed angle rotor, TFT-45.6, titanium, 40×6 ml, 45000 min-1, 238860 ×g (outer), 205250 ×g (inner) 1 SORV11637
Vertical rotor, TV-1665, titanium, 16×6 ml, 65000 min-1, 414360 ×g 1 SORV11672
Fixed angle rotor, T-890, titanium, 8×12,5 ml, 90000 min-1, 692150 ×g 1 SORV11690
Fixed angle rotor, T-1250, titanium, 12×36 ml, 50000 min-1, 301030 ×g 1 SORV11713
Fixed angle rotor, A-621, aluminium, 6×250 ml, 21000 min-1, 66990 ×g 1 SORV11876
Fixed angle rotor, T-8100, titanium, 8×6,5 ml, 100000 min-1, 802010 ×g 1 SORV45621
Continuous flow rotor, TCF-32, 430 ml, 32000 min-1, 102000 ×g (standard core needed for operation) 1 SORV45820
Fixed angle rotor, T-865, titanium, 8×36 ml tubes (25×97 mm), 65000 min-1, 429460 ×g 1 521-1976
Zonal rotor, TZ-28, titanium, 1350 ml, 28000 min-1, 83547 ×g 1 SORV52358
Vertical rotor, TV-860, titanium, 8×35 ml, 60000 min-1, 340600 ×g 1 SORV53289
Swinging bucket rotor, AH-629, aluminium, 6×36 ml, 29000 min-1, 151240 ×g 1 521-1623
Swinging bucket rotor, AH-629, aluminium, 6×17 ml, 29000 min-1, 155850 ×g 1 SORV54285
Swinging bucket rotor AH-629, aluminium, 6×5 ml, 50000 min-1, 296010 ×g 1 SORV54294
Swinging-bucket rotor TH-641, titanium, 6×13,2 ml, 41000 min-1, 287660 ×g 1 SORV54295
Fixed angle rotor, TFT-80,4 , titanium, 10×4,4 ml, 80000 min-1, 468250 ×g 1 SORV54356
Fixed angle rotor TFT-80.2, titanium, 12×2 ml, 80000 min-1, 429640 ×g 1 SORV54456
Vertical rotor, StepSaver™ 70 V6, titanium, 8×6 ml, 70000 min-1, 467420 ×g 1 SORV79240
Vertical rotor, StepSaver™ 65 V13, titanium, 8×13,5 ml, 65000 min-1, 400670 ×g 1 SORV79243
Vertical rotor, StepSaver™ 50 V39, titanium, 8×39 ml, 50000 min-1, 241830 ×g 1 SORV79245
Vertical rotor, StepSaver™ 70 V6, titanium, 8×2 ml, 70000 min-1, 467420 ×g 1 SORV79274
Vertical rotor, StepSaver™ 50 V39, titanium, 8×13,5 ml, 50000 min-1, 228700 ×g 1 SORV79277
Vertical rotor, StepSaver™ 70 V6, titanium, 8×5,1 ml, 70000 min-1, 467420 ×g 1 SORV79280
Vertical rotor, StepSaver™ 65 V13, titanium, quick-seal cartridge set, 8×13,5 ml, 65000 min-1, 400671 ×g 1 SORV79285
Vertical rotor, StepSaver™ 65 V13, titanium, 8×6 ml, 65000 min-1, 393110 ×g 1 SORV79290
Swinging bucket rotor, SureSpin™ 630/17, titanium, 6×17 ml, 30000 min-1, 166880 ×g 1 SORV79363
Swinging bucket rotor, SureSpin™ 630/36, titanium, 6×36 ml, 30000 min-1, 166880 ×g 1 SORV79368
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory. Enter 'x' if unknown for VWR Chemicals

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping